Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 4 | 511 | ||
NM_001024960.2 | NP_001020131.1 | 4 | 366 | |||
NM_001198780.1 | NP_001185709.1 | 4 | 365 | |||
NM_005718.4 | NP_005709.1 | 4 | 877 | |||
GenBank mRNA | AF006087.1 | - | 4 | 318 | ||
AF019888.1 | - | 3 | 303 | |||
AW408422.1 | - | 4 | 324 | |||
BC012596.2 | - | 3 | 300 | |||
BG938423.1 | - | 4 | 349 | |||
BI768875.1 | - | 4 | 490 | |||
BM045872.1 | - | 4 | 376 | |||
BQ688329.1 | - | 4 | 449 | |||
BX419672.2 | - | 4 | 365 | |||
DA226584.1 | - | 4 | 366 | |||
DB060690.1 | - | 4 | 511 |
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 4 | 877 | ||
GenBank mRNA | BC025289.1 | - | 3 | 311 |