Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277140.1 | NP_001264069.1 | 7 | 694 | ||
NM_005721.4 | NP_005712.1 | 7 | 903 | |||
NR_102318.1 | 6 | 778 | ||||
GenBank mRNA | AB209353.1 | - | 5 | 412 | ||
AF006083.1 | - | 7 | 729 | |||
AF127773.1 | - | 7 | 772 | |||
AK300030.1 | - | 6 | 602 | |||
AK300224.1 | - | 6 | 524 | |||
AK302152.1 | - | 7 | 648 | |||
AK302359.1 | - | 7 | 682 | |||
AK312659.1 | - | 7 | 703 | |||
AK316213.1 | - | 7 | 725 | |||
AK316396.1 | - | 7 | 694 | |||
BC044590.1 | - | 7 | 877 | 5261640 |