Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005727.3 | NP_005718.2 | 4, 5 | 734 | ||
XM_011540460.2 | XP_011538762.1 | 4, 5 | 708 | |||
GenBank mRNA | AF054838.1 | - | 2, 3 | 260 | ||
AF065388.1 | - | 3, 4 | 381 | |||
AF133425.1 | - | 3, 4 | 383 | |||
AK313774.1 | - | 4, 5 | 724 | |||
BC007290.1 | - | 3, 4 | 387 | 3349295 | ||
BC013404.2 | - | 3, 4 | 385 | 2959379 | ||
BM908049.1 | - | 4, 5 | 737 |