Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005727.3 | NP_005718.2 | 9 | 1182 | ||
GenBank mRNA | AF054838.1 | - | 7 | 708 | ||
AF065388.1 | - | 8 | 829 | |||
AF133425.1 | - | 8 | 831 | |||
AK313774.1 | - | 9 | 1172 | |||
BC007290.1 | - | 8 | 835 | 3349295 | ||
BC013404.2 | - | 8 | 833 | 2959379 |