Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005733.2 | NP_005724.1 | 12, 13 | 2008 | ||
GenBank mRNA | AF070672.1 | - | 11, 12 | 1539 | ||
AF153329.1 | - | 12, 13 | 1546 | |||
AK024804.1 | - | 1, 2 | 65 | |||
AK025790.1 | - | 12, 13 | 1567 | |||
AK296879.1 | - | 13, 14 | 1522 | |||
BC012999.2 | - | 12, 13 | 1546 | 3604291 |