Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005740.2 | NP_005731.1 | 3 | 321 | ||
GenBank mRNA | AK290566.1 | - | 3 | 274 | ||
AL035366.1 | - | 3 | 314 | |||
AY453399.1 | - | 112 | ||||
BC002968.1 | - | 3 | 288 | 3543962 | ||
BT009847.1 | - | 2 | 93 | |||
BX473356.1 | - | 3 | 326 | |||
CR456487.1 | - | 2 | 178 | |||
CR542177.1 | - | 2 | 93 | |||
CR542195.1 | - | 2 | 93 |