Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015045.2 | NP_001015045.1 | 15 | 2000 | ||
NM_001265578.1 | NP_001252507.1 | 14 | 1958 | |||
NM_001265580.1 | NP_001252509.1 | 13 | 1916 | |||
NM_014883.3 | NP_055698.2 | 21 | 2852 | |||
XM_005262681.2 | XP_005262738.1 | 20 | 2810 | |||
XM_005262682.2 | XP_005262739.1 | 21 | 2951 | |||
XM_005262684.3 | XP_005262741.1 | 20 | 2631 | |||
XM_006714057.3 | XP_006714120.1 | 18 | 2127 | |||
XM_011531516.1 | XP_011529818.1 | 22 | 2873 | |||
XM_011531517.1 | XP_011529819.1 | 20 | 2768 | |||
XM_011531518.1 | XP_011529820.1 | 22 | 3002 | |||
XM_011531519.2 | XP_011529821.1 | 20 | 2561 | |||
XM_017007625.1 | XP_016863114.1 | 20 | 2471 | |||
XM_017007626.1 | XP_016863115.1 | 18 | 2105 | |||
XM_017007627.1 | XP_016863116.1 | 20 | 2696 | |||
XM_017007629.1 | XP_016863118.1 | 21 | 4583 | |||
XM_017007630.1 | XP_016863119.1 | 19 | 2496 | |||
XM_017007631.1 | XP_016863120.1 | 20 | 4541 | |||
XM_017007632.1 | XP_016863121.1 | 18 | 2435 | |||
XM_017007633.1 | XP_016863122.1 | 20 | 4499 | |||
GenBank mRNA | AK027138.1 | - | 15 | 1840 | ||
AK296932.1 | - | 13 | 1785 | |||
AK298262.1 | - | 1 | 73 | |||
BC086875.1 | - | 14 | 1799 | 30708091 |