Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242826.1 | NP_001229755.1 | 5 | 779 | ||
NM_005785.3 | NP_005776.1 | 5 | 840 | |||
NM_194358.2 | NP_919339.1 | 6 | 983 | |||
NM_194359.2 | NP_919340.1 | 5 | 735 | |||
NR_040053.1 | 6 | 983 | ||||
XM_005268561.4 | XP_005268618.1 | 6 | 891 | |||
XM_011537734.2 | XP_011536036.1 | 6 | 897 | |||
XM_011537735.2 | XP_011536037.1 | 6 | 814 | |||
GenBank mRNA | AF077599.1 | - | 4 | 673 | ||
AK300752.1 | - | 6 | 841 | |||
AK304576.1 | - | 6 | 680 | |||
AK314811.1 | - | 5 | 699 | |||
BC024284.1 | - | 6 | 961 | |||
BC032637.1 | - | 4 | 688 | 5496849 | ||
BC036267.1 | - | 5 | 784 | |||
BC095418.1 | - | 6 | 918 | |||
BU170070.1 | - | 5 | 716 |