Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321532.1 | NP_001308461.1 | 3 | 920 | ||
NM_005800.4 | NP_005791.3 | 4 | 1073 | |||
XM_005266213.3 | XP_005266270.1 | 5 | 1431 | |||
XM_006719751.3 | XP_006719814.1 | 4 | 1351 | |||
XM_017020314.1 | XP_016875803.1 | 4 | 1369 | |||
XM_017020315.1 | XP_016875804.1 | 3 | 740 | |||
XM_017020316.1 | XP_016875805.1 | 4 | 1264 | |||
GenBank mRNA | AK289826.1 | - | 4 | 829 | ||
AL832776.1 | - | 3 | 639 | |||
BC036027.1 | - | 4 | 845 | 5271146 | ||
BC038103.2 | - | 4 | 983 | 4821753 | ||
X59131.1 | - | 4 | 818 |