Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDSPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008392.1 | NP_001008393.1 | 7 | 709 | ||
NM_005808.2 | NP_005799.2 | 6 | 676 | |||
XM_017005519.1 | XP_016861008.1 | 7 | 735 | |||
XM_017005520.1 | XP_016861009.1 | 7 | 2053 | |||
GenBank mRNA | AJ575644.1 | - | 6 | 676 | ||
AJ575645.1 | - | 7 | 709 | |||
AY279532.1 | - | 6 | 654 | |||
AY364238.1 | - | 6 | 654 | |||
D88153.1 | - | 7 | 1026 |