Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005876.4 | NP_005867.3 | 14 | 3802 | ||
XM_005246237.2 | XP_005246294.1 | 16 | 3530 | |||
XM_005246239.2 | XP_005246296.1 | 16 | 3345 | |||
XM_005246240.2 | XP_005246297.1 | 9 | 1547 | |||
XM_005246241.1 | XP_005246298.1 | 9 | 1441 | |||
XM_005246242.4 | XP_005246299.1 | 9 | 1566 | |||
XM_006712189.3 | XP_006712252.1 | 15 | 3535 | |||
XM_006712193.3 | XP_006712256.1 | 9 | 4666 | |||
XM_011510479.2 | XP_011508781.1 | 15 | 3968 | |||
XM_011510483.2 | XP_011508785.2 | 15 | 3565 | |||
XM_017003157.1 | XP_016858646.1 | 15 | 3565 | |||
XM_017003158.1 | XP_016858647.1 | 11 | 3141 | |||
XM_017003159.1 | XP_016858648.1 | 12 | 3324 | |||
GenBank mRNA | AB037718.1 | - | 5 | 859 | ||
AK098307.1 | - | 1 | 293 | |||
AY603755.1 | - | 13 | 3415 |