Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005880.3 | NP_005871.1 | 9 | 1308 | ||
GenBank mRNA | AF088046.1 | - | 3 | 284 | ||
AJ001309.1 | - | 9 | 1218 | |||
AK313031.1 | - | 9 | 1256 | |||
BC013044.2 | - | 9 | 1260 | 3505399 | ||
BC015809.2 | - | 9 | 1261 | 3922477 | ||
Y13350.1 | - | 7 | 903 |