Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 3 | 467 | ||
NM_001297431.1 | NP_001284360.1 | 3 | 467 | |||
NM_001297432.1 | NP_001284361.1 | 3 | 605 | |||
NM_001297433.1 | NP_001284362.1 | 3 | 541 | |||
NM_005882.4 | NP_005873.2 | 3 | 467 | |||
NR_123716.1 | 3 | 467 | ||||
XM_006713849.3 | XP_006713912.1 | 3 | 427 | |||
XM_006713850.2 | XP_006713913.1 | 3 | 600 | |||
XM_011513376.1 | XP_011511678.1 | 4 | 587 | |||
XM_011513377.1 | XP_011511679.1 | 3 | 505 | |||
XM_017007636.1 | XP_016863125.1 | 4 | 587 | |||
XM_017007637.1 | XP_016863126.1 | 3 | 504 | |||
GenBank mRNA | AF084928.1 | - | 4 | 415 | ||
AK001088.1 | - | 3 | 427 | |||
AK022515.1 | - | 3 | 436 | |||
AK022586.1 | - | 3 | 427 | |||
AK094253.1 | - | 3 | 397 | |||
AK298944.1 | - | 3 | 521 | |||
AK300572.1 | - | 4 | 364 | |||
AK301155.1 | - | 3 | 435 | |||
AK308901.1 | - | 3 | 411 | |||
AY189687.1 | - | 3 | 380 | |||
AY236486.1 | - | 3 | 412 | |||
BC001225.1 | - | 3 | 434 | 3533990 | ||
BC006470.1 | - | 3 | 390 | |||
BT006957.1 | - | 3 | 371 | |||
DC299400.1 | - | 3 | 544 |