Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 8 | 1076 | ||
NM_001297430.1 | NP_001284359.1 | 7 | 872 | |||
NM_001297431.1 | NP_001284360.1 | 6 | 872 | |||
NM_001297432.1 | NP_001284361.1 | 8 | 1214 | |||
NM_001297433.1 | NP_001284362.1 | 8 | 1150 | |||
NM_005882.4 | NP_005873.2 | 7 | 953 | |||
NR_123716.1 | 6 | 876 | ||||
XM_006713849.3 | XP_006713912.1 | 8 | 1036 | |||
XM_006713850.2 | XP_006713913.1 | 8 | 1209 | |||
XM_011513376.1 | XP_011511678.1 | 9 | 1196 | |||
XM_011513377.1 | XP_011511679.1 | 8 | 1114 | |||
XM_017007636.1 | XP_016863125.1 | 9 | 1196 | |||
XM_017007637.1 | XP_016863126.1 | 8 | 1113 | |||
GenBank mRNA | AB208907.1 | - | 3 | 3776 | ||
AF084928.1 | - | 8 | 913 | |||
AK001088.1 | - | 8 | 1036 | |||
AK022515.1 | - | 6 | 841 | |||
AK022586.1 | - | 7 | 913 | |||
AK091951.1 | - | 3 | 947 | |||
AK094253.1 | - | 5 | 697 | |||
AK125501.1 | - | 1 | 872 | |||
AK127875.1 | - | 4 | 1304 | |||
AK128302.1 | - | 2 | 1980 | |||
AK296174.1 | - | 7 | 829 | |||
AK298944.1 | - | 8 | 1130 | |||
AK301155.1 | - | 8 | 1044 | |||
AY189687.1 | - | 8 | 989 | |||
AY236486.1 | - | 6 | 821 | |||
BC001225.1 | - | 8 | 1043 | 3533990 | ||
BC006470.1 | - | 8 | 999 | |||
BT006957.1 | - | 8 | 980 |