Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006059.3 | NP_006050.3 | 12 | 2104 | ||
XM_006716921.2 | XP_006716984.1 | 12 | 2047 | |||
XM_011518121.1 | XP_011516423.1 | 12 | 2047 | |||
GenBank mRNA | AB208887.1 | - | 11 | 1420 | ||
AF041835.1 | - | 12 | 2103 |