Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011529130.2 | XP_011527432.1 | 1 | 186 | ||
XM_011529134.2 | XP_011527436.1 | 1 | 186 | |||
XM_017027577.1 | XP_016883066.1 | 1 | 533 | |||
XM_017027578.1 | XP_016883067.1 | 1 | 538 | |||
XM_017027579.1 | XP_016883068.1 | 1 | 539 | |||
XM_017027580.1 | XP_016883069.1 | 1 | 539 | |||
XM_017027581.1 | XP_016883070.1 | 1 | 536 | |||
XM_017027582.1 | XP_016883071.1 | 1 | 549 | |||
XM_017027583.1 | XP_016883072.1 | 1 | 533 | |||
XM_017027584.1 | XP_016883073.1 | 1 | 533 | |||
XM_017027585.1 | XP_016883074.1 | 1 | 530 | |||
GenBank mRNA | AK289866.1 | - | 1 | 101 | ||
AK313517.1 | - | 1 | 101 | |||
BC025286.1 | - | 1 | 88 | |||
BC075835.1 | - | 1 | 88 | |||
BC131747.1 | - | 1 | 72 | |||
BM010247.1 | - | 1 | 89 | |||
BX537992.1 | - | 1 | 102 | |||
DA463753.1 | - | 1 | 135 | |||
DA761891.1 | - | 1 | 188 | |||
DA967881.1 | - | 1 | 154 | |||
Y10376.1 | - | 1 | 76 |