Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 16, 17 | 2282 | ||
XM_017024011.1 | XP_016879500.1 | 16, 17 | 2247 | |||
XM_017024012.1 | XP_016879501.1 | 16, 17 | 2351 | |||
XM_017024013.1 | XP_016879502.1 | 16, 17 | 2171 | |||
XM_017024014.1 | XP_016879503.1 | 16, 17 | 2771 | |||
XM_017024015.1 | XP_016879504.1 | 13, 14 | 1794 | |||
GenBank mRNA | AF423307.1 | - | 15, 16 | 2139 | ||
AK057068.1 | - | 16, 17 | 2214 | |||
AK074491.1 | - | 9, 10 | 1091 | |||
AY028899.1 | - | 16, 17 | 2282 | |||
CR627382.1 | - | 15, 16 | 2088 |