Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080283.3 | NP_525022.2 | 26 | 3594 | ||
XM_017024011.1 | XP_016879500.1 | 26 | 3559 | |||
XM_017024012.1 | XP_016879501.1 | 26 | 3663 | |||
XM_017024013.1 | XP_016879502.1 | 26 | 3483 | |||
XM_017024014.1 | XP_016879503.1 | 26 | 4083 | |||
XM_017024015.1 | XP_016879504.1 | 23 | 3106 | |||
GenBank mRNA | AF423307.1 | - | 25 | 3451 | ||
AY028899.1 | - | 26 | 3594 | |||
CR627382.1 | - | 24 | 3286 |