Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015836.3 | NP_056651.1 | 3 | 396 | ||
NM_201263.2 | NP_957715.1 | 3 | 396 | |||
XM_005270350.3 | XP_005270407.1 | 3 | 482 | |||
XM_011540494.2 | XP_011538796.1 | 4 | 465 | |||
XM_011540495.2 | XP_011538797.1 | 2 | 167 | |||
XM_017000038.1 | XP_016855527.1 | 3 | 388 | |||
XM_017000039.1 | XP_016855528.1 | 5 | 651 | |||
XM_017000040.1 | XP_016855529.1 | 3 | 218 | |||
XM_017000041.1 | XP_016855530.1 | 3 | 365 | |||
XM_017000042.1 | XP_016855531.1 | 3 | 388 | |||
GenBank mRNA | AJ242739.1 | - | 3 | 377 | ||
AK223197.1 | - | 3 | 374 | |||
AK296088.1 | - | 3 | 361 | |||
AK296799.1 | - | 3 | 377 | |||
AK299547.1 | - | 3 | 335 | |||
AK300295.1 | - | 3 | 224 | |||
AK313740.1 | - | 3 | 394 | |||
BC021722.1 | - | 3 | 375 | |||
BC039889.1 | - | 3 | 372 | |||
BC044575.1 | - | 3 | 379 | 5287131 | ||
BM834499.1 | - | 3 | 388 |