Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323565.1 | NP_001310494.1 | 1 | 350 | ||
NM_001323566.1 | NP_001310495.1 | 1 | 350 | |||
NM_001801.2 | NP_001792.2 | 1 | 350 | |||
NR_136618.1 | 1 | 350 | ||||
NR_136619.1 | 1 | 350 | ||||
NR_136620.1 | 1 | 350 | ||||
GenBank mRNA | AK130357.1 | - | 1 | 312 | ||
AK314231.1 | - | 1 | 300 | |||
AL534761.3 | - | 1 | 349 | |||
BC024241.2 | - | 1 | 300 | 4797463 | ||
CR536540.1 | - | 1 | 39 | |||
D85777.1 | - | 1 | 268 | |||
DA359016.1 | - | 1 | 300 | |||
DA849625.1 | - | 1 | 299 | |||
Z23010.1 | - | 1 | 39 | |||
Z31357.1 | - | 1 | 293 |