Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 10 | 1411 | ||
NR_033717.1 | 10 | 1411 | ||||
XM_006724458.1 | XP_006724521.1 | 10 | 1375 | |||
XM_017029219.1 | XP_016884708.1 | 10 | 1307 | |||
XM_017029220.1 | XP_016884709.1 | 10 | 1303 | |||
XM_017029221.1 | XP_016884710.1 | 10 | 1445 | |||
XM_017029222.1 | XP_016884711.1 | 4 | 689 | |||
GenBank mRNA | AK299190.1 | - | 4 | 689 | ||
AK303064.1 | - | 9 | 1277 | |||
AK308140.1 | - | 4 | 685 | |||
AK315116.1 | - | 10 | 1375 | |||
BC051913.1 | - | 10 | 1353 | |||
BC064617.1 | - | 10 | 1368 | 5575894 | ||
Y18004.1 | - | 10 | 1345 |