Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256706.1 | NP_001243635.1 | 5 | 603 | ||
NM_001256707.1 | NP_001243636.1 | 6 | 676 | |||
NM_001256708.1 | NP_001243637.1 | 5 | 809 | |||
NM_001256709.1 | NP_001243638.1 | 7 | 744 | |||
NM_001256710.1 | NP_001243639.1 | 6 | 641 | |||
NM_001256711.1 | NP_001243640.1 | 7 | 714 | |||
NM_001256712.1 | NP_001243641.1 | 4 | 471 | |||
NM_001318367.1 | NP_001305296.1 | 5 | 641 | |||
NM_014885.4 | NP_055700.2 | 5 | 592 | |||
NR_046345.1 | 3 | 355 | ||||
XM_011531524.2 | XP_011529826.1 | 6 | 832 | |||
XM_011531531.2 | XP_011529833.1 | 4 | 498 | |||
XM_017007638.1 | XP_016863127.1 | 5 | 619 | |||
XM_017007639.1 | XP_016863128.1 | 6 | 1268 | |||
XM_017007640.1 | XP_016863129.1 | 5 | 660 | |||
XM_017007643.1 | XP_016863132.1 | 6 | 657 | |||
XM_017007644.1 | XP_016863133.1 | 6 | 631 | |||
GenBank mRNA | AB012109.1 | - | 5 | 590 | ||
AF132794.1 | - | 5 | 547 | |||
AL080090.1 | - | 6 | 650 | |||
AV757106.2 | - | 4 | 530 | |||
BC005217.1 | - | 5 | 569 | 3929766 | ||
BG530912.1 | - | 7 | 688 | |||
BI488888.1 | - | 3 | 353 | |||
BI913723.1 | - | 5 | 642 | |||
BU175327.1 | - | 5 | 573 | |||
CV023091.1 | - | 4 | 491 |