Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006097.4 | NP_006088.2 | 2 | 208 | ||
NM_181526.2 | NP_852667.1 | 2 | 208 | |||
GenBank mRNA | AF176042.1 | - | 1 | 144 | ||
AF176043.1 | - | 1 | 144 | |||
AK097235.1 | - | 1 | 3464 | |||
AK130879.1 | - | 2 | 173 | |||
AL697699.1 | - | 2 | 208 | |||
BC002648.2 | - | 2 | 147 | 3608316 | ||
BQ953325.1 | - | 198 | ||||
CR456843.1 | - | 1 | 104 | |||
D82057.1 | - | 2 | 176 | |||
J02854.1 | - | 1 | 146 |
Gene Symbol: SLC2A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286233.1 | NP_001273162.1 | 5 | 775 | ||
NM_001286235.1 | NP_001273164.1 | 3 | 408 | |||
XM_011520562.1 | XP_011518864.1 | 4 | 298 | |||
XM_017018842.1 | XP_016874331.1 | 4 | 542 | |||
XM_017018843.1 | XP_016874332.1 | 4 | 866 | |||
XM_017018846.1 | XP_016874335.1 | 2 | 119 | |||
GenBank mRNA | AK122783.1 | - | 3 | 199 | ||
AK126026.1 | - | 5 | 560 | |||
AY357941.1 | - | 1 | 95 | |||
BG717327.1 | - | 2 | 172 | |||
DB061127.1 | - | 3 | 193 | |||
DB062960.1 | - | 4 | 298 |