Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025107.2 | NP_001020278.1 | 6 | 2326 | ||
NM_001111.4 | NP_001102.2 | 6 | 2457 | |||
NM_001193495.1 | NP_001180424.1 | 6 | 2495 | |||
NM_015840.3 | NP_056655.2 | 6 | 2457 | |||
NM_015841.3 | NP_056656.2 | 6 | 2400 | |||
XM_006711109.2 | XP_006711172.1 | 6 | 2263 | |||
XM_006711111.3 | XP_006711174.1 | 7 | 2385 | |||
XM_006711112.2 | XP_006711175.1 | 7 | 2087 | |||
XM_006711113.2 | XP_006711176.1 | 7 | 2161 | |||
XM_011509060.1 | XP_011507362.1 | 6 | 2359 | |||
XM_011509061.2 | XP_011507363.2 | 6 | 2326 | |||
XM_011509062.1 | XP_011507364.1 | 6 | 2286 | |||
XM_017000037.1 | XP_016855526.1 | 7 | 2254 | |||
GenBank mRNA | AB209891.1 | - | 6 | 2270 | ||
AK304153.1 | - | 6 | 2411 | |||
BC038227.1 | - | 6 | 2384 | 5497548 | ||
BU430287.1 | - | 3 | 334 | |||
BX538232.1 | - | 6 | 2310 | |||
BX640741.1 | - | 6 | 2308 | |||
DR158819.1 | - | 5 | 693 | |||
KJ544856.1 | - | 5 | 2215 | |||
U10439.1 | - | 6 | 2369 | |||
U18121.1 | - | 6 | 2262 | |||
X79448.1 | - | 6 | 2495 | |||
X79449.1 | - | 6 | 2417 | |||
X98559.1 | - | 5 | 2200 |