Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006103.3 | NP_006094.3 | 2, 3 | 241 | ||
GenBank mRNA | AK225528.1 | - | 2, 3 | 240 | ||
AK311966.1 | - | 2, 3 | 242 | |||
AY212888.1 | - | 2, 3 | 250 | |||
BC039816.1 | - | 2, 3 | 218 | |||
BC046106.1 | - | 2, 3 | 217 | 5186932 | ||
BI759775.1 | - | 2, 3 | 269 | |||
CR456977.1 | - | 2, 3 | 213 |