Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271665.1 | NP_001258594.1 | 4 | 840 | ||
NM_014886.4 | NP_055701.1 | 4 | 840 | |||
NR_073403.1 | 4 | 840 | ||||
XM_011543098.1 | XP_011541400.1 | 4 | 518 | |||
XM_017008952.1 | XP_016864441.1 | 4 | 518 | |||
XM_017008953.1 | XP_016864442.1 | 4 | 590 | |||
GenBank mRNA | AF077615.1 | - | 4 | 554 | ||
AF372458.1 | - | 4 | 565 | |||
AJ012409.1 | - | 4 | 552 | |||
AK313784.1 | - | 4 | 583 | |||
AY443419.1 | - | 3 | 470 | |||
BC005288.1 | - | 4 | 583 | 3683720 | ||
BQ775601.1 | - | 4 | 584 | |||
CB052084.1 | - | 2 | 205 |