Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014886.4 | NP_055701.1 | 5 | 997 | ||
NR_073403.1 | 6 | 1106 | ||||
XM_011543098.1 | XP_011541400.1 | 5 | 675 | |||
GenBank mRNA | AA830219.1 | - | 2 | 401 | ||
AA921837.1 | - | 2 | 288 | |||
AF077615.1 | - | 5 | 711 | |||
AF372458.1 | - | 5 | 722 | |||
AJ012409.1 | - | 5 | 709 | |||
AK313784.1 | - | 5 | 740 | |||
AY443419.1 | - | 4 | 627 | |||
BC005288.1 | - | 5 | 740 | 3683720 | ||
BQ775601.1 | - | 2 | 317 |