Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016172.2 | NP_057256.2 | 10 | 1412 | ||
GenBank mRNA | AF038200.1 | - | 9 | 977 | ||
AF068195.1 | - | 9 | 1039 | |||
AF176796.1 | - | 10 | 1382 | |||
BC004967.1 | - | 10 | 1412 | 3535682 | ||
BC011822.1 | - | 10 | 1238 | 3688526 | ||
BX647463.1 | - | 3 | 5465 |