Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242597.1 | NP_001229526.1 | 4 | 621 | ||
NM_032635.3 | NP_116024.1 | 5 | 550 | |||
GenBank mRNA | AW130399.1 | - | 3 | 357 | ||
AW473288.1 | - | 3 | 343 | |||
BC001118.2 | - | 5 | 507 | 2989840 | ||
BG697070.1 | - | 4 | 510 | |||
BM423809.1 | - | 5 | 550 | |||
Y18007.1 | - | 5 | 518 |