Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MERTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006343.2 | NP_006334.2 | 17 | 2402 | ||
XM_005263565.4 | XP_005263622.1 | 17 | 2537 | |||
XM_011510490.2 | XP_011508792.1 | 17 | 2437 | |||
XM_017003164.1 | XP_016858653.1 | 17 | 2407 | |||
XM_017003165.1 | XP_016858654.1 | 18 | 2506 | |||
GenBank mRNA | AK294196.1 | - | 9 | 848 | ||
AK308695.1 | - | 9 | 850 | |||
AK316584.1 | - | 17 | 2409 | |||
BC114917.1 | - | 17 | 2116 | 40068588 | ||
BC114918.1 | - | 17 | 2116 | 40012335 | ||
U08023.1 | - | 17 | 2417 |