Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006344.2 | NP_006335.2 | 4 | 509 | ||
NM_182906.2 | NP_878910.1 | 4 | 509 | |||
XM_005256412.1 | XP_005256469.1 | 4 | 535 | |||
XM_011523613.1 | XP_011521915.1 | 4 | 535 | |||
XM_011523614.2 | XP_011521916.1 | 5 | 601 | |||
XM_011523615.1 | XP_011521917.1 | 4 | 535 | |||
GenBank mRNA | AK291975.1 | - | 4 | 472 | ||
AK292363.1 | - | 4 | 476 | |||
AL546075.3 | - | 4 | 458 | |||
BC027858.1 | - | 4 | 456 | 5225381 | ||
BC039011.1 | - | 4 | 457 | 5751735 | ||
BI910082.1 | - | 4 | 509 | |||
D50532.1 | - | 4 | 413 |