Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006351.3 | NP_006342.2 | 9, 10 | 1155 | ||
GenBank mRNA | AF026030.1 | - | 9, 10 | 972 | ||
AF041254.1 | - | 9, 10 | 1013 | |||
AK223062.1 | - | 9, 10 | 998 | |||
AK289634.1 | - | 9, 10 | 1155 | |||
AK309415.1 | - | 10, 11 | 1106 | |||
BC033628.1 | - | 9, 10 | 985 | 5248163 |