Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042537.1 | NP_001036002.1 | 1, 2 | 392 | ||
NM_001177651.1 | NP_001171122.1 | 2, 3 | 270 | |||
NM_006359.2 | NP_006350.1 | 1, 2 | 392 | |||
XM_006724726.3 | XP_006724789.1 | 2, 3 | 386 | |||
XM_011531243.2 | XP_011529545.1 | 2, 3 | 246 | |||
XM_017029223.1 | XP_016884712.1 | 2, 3 | 403 | |||
XM_017029224.1 | XP_016884713.1 | 2, 3 | 533 | |||
XM_017029225.1 | XP_016884714.1 | 2, 3 | 266 | |||
GenBank mRNA | AB074255.1 | - | 1, 2 | 316 | ||
AF030409.1 | - | 1, 2 | 351 | |||
AK289775.1 | - | 1, 2 | 316 | |||
AK296444.1 | - | 1, 2 | 55 | |||
AK300475.1 | - | 2, 3 | 261 | |||
AK314562.1 | - | 1, 2 | 384 | |||
AK316255.1 | - | 2, 3 | 261 | |||
BC035029.1 | - | 1, 2 | 354 | 5264865 | ||
BC049169.1 | - | 1, 2 | 316 | 5312949 | ||
D87743.1 | - | 1, 2 | 307 | |||
DB483081.1 | - | 1, 2 | 392 | |||
DC308430.1 | - | 2, 3 | 271 |