Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032108.3 | NP_115484.2 | 6 | 652 | ||
XM_011527639.2 | XP_011525941.1 | 6 | 701 | |||
XM_011527640.2 | XP_011525942.1 | 6 | 444 | |||
XM_011527641.1 | XP_011525943.1 | 6 | 701 | |||
GenBank mRNA | AB022433.1 | - | 5 | 428 | ||
AB209278.1 | - | 5 | 414 | |||
AF216389.1 | - | 5 | 393 | |||
AF293363.1 | - | 5 | 393 | |||
AK300037.1 | - | 6 | 652 | |||
AY358939.1 | - | 5 | 408 | |||
BC142617.1 | - | 6 | 595 | 40147342 | ||
FR839673.1 | - | 5 | 390 |