Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105538.1 | NP_001099008.1 | 6 | 667 | ||
NM_014520.3 | NP_055335.2 | 6 | 667 | |||
XM_011523616.2 | XP_011521918.1 | 6 | 648 | |||
GenBank mRNA | AF147709.1 | - | 6 | 630 | ||
AK301622.1 | - | 5 | 740 | |||
AK303150.1 | - | 6 | 635 | |||
AK307948.1 | - | 6 | 667 | |||
AL136595.1 | - | 5 | 375 | |||
BC050546.1 | - | 6 | 610 | 5495483 |