Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014842.2 | NP_001014842.1 | 3 | 931 | ||
NM_001289006.1 | NP_001275935.1 | 3 | 725 | |||
NM_006405.6 | NP_006396.2 | 3 | 931 | |||
GenBank mRNA | AK292110.1 | - | 3 | 620 | ||
BC010856.2 | - | 2 | 540 | 3899246 | ||
BX161390.1 | - | 3 | 851 | |||
BX161494.1 | - | 3 | 813 | |||
U94831.1 | - | 2 | 538 |