Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006407.3 | NP_006398.1 | 2 | 325 | ||
GenBank mRNA | AB014765.1 | - | 2 | 231 | ||
AB052638.1 | - | 2 | 223 | |||
AB097051.1 | - | 2 | 306 | |||
AF070523.1 | - | 2 | 331 | |||
AF115965.1 | - | 2 | 316 | |||
AF125530.1 | - | 2 | 317 | |||
AF161476.1 | - | 2 | 306 | |||
AK223566.1 | - | 2 | 229 | |||
AK301637.1 | - | 2 | 280 | |||
AK312731.1 | - | 2 | 228 | |||
AY102608.1 | - | 2 | 216 | |||
BC005143.2 | - | 2 | 248 | 3510436 | ||
BC020797.1 | - | 2 | 242 | 4134658 | ||
BG483337.1 | - | 326 |