Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190996.1 | NP_001177925.1 | 8, 9 | 1144 | ||
NM_006409.3 | NP_006400.2 | 8, 9 | 1146 | |||
GenBank mRNA | AB209059.1 | - | 8, 9 | 1079 | ||
AK223015.1 | - | 7, 8 | 970 | |||
BC039594.1 | - | 8, 9 | 1109 | 6018804 | ||
BC047889.1 | - | 8, 9 | 1079 | 6473050 | ||
BC054027.1 | - | 8, 9 | 1078 | 6514889 | ||
EU668326.1 | - | 8, 9 | 1132 | |||
GQ472234.1 | - | 7, 8 | 970 | |||
Y08999.1 | - | 7, 8 | 1018 |