Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTATIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098520.1 | NP_001091990.1 | 3 | 411 | ||
NM_001098521.1 | NP_001091991.1 | 3 | 339 | |||
NM_001098522.1 | NP_001091992.1 | 2 | 621 | |||
NM_001098523.1 | NP_001091993.1 | 2 | 337 | |||
NM_006410.4 | NP_006401.3 | 3 | 306 | |||
GenBank mRNA | AF039103.1 | - | 2 | 252 | ||
AF092095.1 | - | 2 | 338 | |||
AK222969.1 | - | 2 | 295 | |||
AK223010.1 | - | 2 | 329 | |||
AK292092.1 | - | 3 | 315 | |||
AW963626.1 | - | 2 | 256 | |||
BC002439.2 | - | 3 | 352 | 3346984 | ||
BC015358.2 | - | 2 | 317 | 3455757 | ||
BF681178.1 | - | 305 | ||||
BP382529.1 | - | 3 | 411 | |||
CR456821.1 | - | 2 | 240 | |||
DB450790.1 | - | 3 | 338 | |||
U69161.1 | - | 2 | 338 |