Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012727.1 | NP_001012745.1 | 4 | 652 | ||
NM_006412.3 | NP_006403.2 | 5 | 748 | |||
GenBank mRNA | AF000237.1 | - | 5 | 712 | ||
AF011374.1 | - | 5 | 706 | |||
BC000026.2 | - | 5 | 691 | 3507150 | ||
BC004529.2 | - | 4 | 586 | 3347049 | ||
BC007269.1 | - | 3 | 323 | |||
BC019292.1 | - | 5 | 700 | 2823580 | ||
U56418.2 | - | 5 | 685 |