Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIVA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006427.3 | NP_006418.2 | 1, 2 | 195 | ||
XM_011536360.2 | XP_011534662.1 | 1, 2 | 908 | |||
GenBank mRNA | AK128704.1 | - | 1, 2 | 126 | ||
AK223332.1 | - | 1, 2 | 181 | |||
AK300415.1 | - | 1, 2 | 144 | |||
AK316457.1 | - | 1, 2 | 126 | |||
AK316550.1 | - | 1, 2 | 181 | |||
BU195069.1 | - | 1, 2 | 195 | |||
CK001877.1 | - | 1, 2 | 189 |