Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006470.3 | NP_006461.3 | 4 | 799 | ||
GenBank mRNA | AF096870.1 | - | 1 | 469 | ||
AK002035.1 | - | 1 | 265 | |||
AK314557.1 | - | 1 | 279 | |||
BC001564.1 | - | 1 | 129 | |||
BC053514.1 | - | 4 | 799 | 6263729 | ||
BC067096.1 | - | 4 | 799 | 6088764 | ||
CR457136.1 | - | 1 | 242 |