Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF6L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006473.3 | NP_006464.1 | 3 | 384 | ||
XM_005273714.1 | XP_005273771.1 | 3 | 276 | |||
GenBank mRNA | AF069735.1 | - | 2 | 183 | ||
AK289499.1 | - | 3 | 294 | |||
AK301145.1 | - | 3 | 284 | |||
AK310054.1 | - | 3 | 285 | |||
AK314330.1 | - | 3 | 270 | |||
BC008785.2 | - | 3 | 284 | |||
DC311213.1 | - | 3 | 384 |