Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006635.2 | NP_001006636.1 | 5 | 614 | ||
NM_001319097.1 | NP_001306026.1 | 4 | 410 | |||
NM_001319098.1 | NP_001306027.1 | 4 | 410 | |||
NM_006554.4 | NP_006545.1 | 4 | 410 | |||
XM_011510497.2 | XP_011508799.1 | 5 | 507 | |||
GenBank mRNA | AF053551.1 | - | 4 | 383 | ||
AK001287.1 | - | 4 | 362 | |||
AK289359.1 | - | 4 | 361 | |||
AK303564.1 | - | 5 | 539 | |||
AK315918.1 | - | 5 | 550 | |||
BC017271.2 | - | 5 | 544 | 4635884 | ||
BC067831.1 | - | 4 | 389 | 5298149 | ||
BC088359.1 | - | 4 | 345 | 30378040 | ||
BG699784.1 | - | 5 | 617 | |||
BU165147.1 | - | 4 | 341 | |||
CX165048.1 | - | 4 | 303 |