Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006573.4 | NP_006564.1 | 3 | 727 | ||
GenBank mRNA | AF116456.1 | - | 3 | 578 | ||
AF132600.1 | - | 3 | 596 | |||
AF134715.1 | - | 3 | 604 | |||
AF136293.1 | - | 3 | 460 | |||
AF186114.1 | - | 3 | 617 | |||
AK309629.1 | - | 4 | 497 | |||
AY129225.1 | - | 3 | 460 | |||
AY129226.1 | - | 3 | 229 | |||
AY129227.1 | - | 2 | 127 | |||
AY129228.1 | - | 2 | 79 | |||
AY358881.1 | - | 3 | 629 | |||
BC020674.1 | - | 3 | 639 | 4719315 | ||
CR541818.1 | - | 3 | 460 | |||
HM636064.1 | - | 3 | 460 |