Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006579.2 | NP_006570.1 | 3, 4 | 543 | ||
GenBank mRNA | BC001549.1 | - | 3, 4 | 473 | 3463689 | |
BC001572.1 | - | 3, 4 | 475 | 3453423 | ||
BC046501.1 | - | 3, 4 | 517 | 5106272 | ||
CA488777.1 | - | 3, 4 | 523 | |||
CR456815.1 | - | 2, 3 | 338 | |||
CR542094.1 | - | 2, 3 | 338 | |||
KJ921656.1 | - | 2, 3 | 338 | |||
Z37986.1 | - | 3, 4 | 449 |