Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006598.2 | NP_006589.2 | 2 | 257 | ||
XM_005248231.3 | XP_005248288.1 | 2 | 262 | |||
XM_011513939.2 | XP_011512241.1 | 2 | 351 | |||
XM_011513940.2 | XP_011512242.1 | 2 | 186 | |||
XM_011513941.2 | XP_011512243.2 | 2 | 281 | |||
XM_017008958.1 | XP_016864447.1 | 2 | 561 | |||
XM_017008959.1 | XP_016864448.1 | 2 | 213 | |||
GenBank mRNA | AF105365.1 | - | 2 | 195 | ||
AK302856.1 | - | 2 | 235 | |||
BC007760.2 | - | 2 | 237 | |||
BC018982.2 | - | 2 | 237 | |||
BC098390.1 | - | 2 | 235 | 5265743 | ||
DB451347.1 | - | 2 | 257 |