Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042749.2 | NP_001036214.1 | 9 | 1162 | ||
NM_001042750.1 | NP_001036215.1 | 9 | 1006 | |||
NM_001042751.1 | NP_001036216.1 | 9 | 1006 | |||
NM_001282418.1 | NP_001269347.1 | 9 | 985 | |||
NM_006603.4 | NP_006594.3 | 8 | 941 | |||
XM_005262357.2 | XP_005262414.1 | 8 | 1031 | |||
XM_005262358.2 | XP_005262415.1 | 9 | 968 | |||
XM_005262359.3 | XP_005262416.1 | 8 | 921 | |||
XM_005262360.2 | XP_005262417.1 | 8 | 850 | |||
XM_005262361.2 | XP_005262418.1 | 8 | 978 | |||
XM_006724727.1 | XP_006724790.1 | 10 | 1110 | |||
XM_006724728.3 | XP_006724791.1 | 9 | 1044 | |||
XM_011531253.2 | XP_011529555.1 | 8 | 903 | |||
XM_017029232.1 | XP_016884721.1 | 9 | 968 | |||
XM_017029233.1 | XP_016884722.1 | 8 | 850 | |||
XM_017029234.1 | XP_016884723.1 | 8 | 903 | |||
XM_017029235.1 | XP_016884724.1 | 9 | 1049 | |||
GenBank mRNA | AK098737.1 | - | 8 | 985 | ||
AK313559.1 | - | 8 | 833 | |||
AL831939.2 | - | 8 | 909 | |||
BC001765.1 | - | 8 | 890 | 3352347 | ||
BX640970.1 | - | 9 | 1017 | |||
BX641002.1 | - | 8 | 910 | |||
BX641003.1 | - | 9 | 975 | |||
CR627425.1 | - | 9 | 984 | |||
CR749433.1 | - | 9 | 975 |