Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006609.4 | NP_006600.3 | 5, 6 | 476 | ||
GenBank mRNA | AB208963.1 | - | 4, 5 | 382 | ||
AF111105.1 | - | 5, 6 | 477 | |||
AF239798.1 | - | 4, 5 | 407 | |||
AK315007.1 | - | 6, 7 | 660 | |||
BC136293.1 | - | 5, 6 | 445 | 9020280 |