Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005716.3 | NP_005707.1 | 7, 8 | 1056 | ||
NM_202468.2 | NP_974197.1 | 6, 7 | 1000 | |||
NM_202469.2 | NP_974198.1 | 5, 6 | 682 | |||
NM_202470.2 | NP_974199.1 | 5, 6 | 912 | |||
NM_202494.2 | NP_974223.1 | 4, 5 | 594 | |||
XM_017026147.1 | XP_016881636.1 | 6, 7 | 947 | |||
XM_017026148.1 | XP_016881637.1 | 6, 7 | 711 | |||
GenBank mRNA | AF028824.1 | - | 4, 5 | 540 | ||
AF089816.1 | - | 5, 6 | 856 | |||
AK022585.1 | - | 4, 5 | 771 | |||
AK093820.1 | - | 1, 2 | 1186 | |||
AK290252.1 | - | 6, 7 | 959 | |||
AK290948.1 | - | 5, 6 | 648 | |||
BC000410.2 | - | 7, 8 | 1028 | 2960992 | ||
BC004226.2 | - | 4, 5 | 671 | |||
BC012810.2 | - | 7, 8 | 1028 | 2960992 | ||
BC016169.1 | - | 7, 8 | 1028 | 3530664 | ||
CR986168.1 | - | 4, 5 | 559 |